missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LEDGF Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3468
This item is not returnable.
View return policy
Description
LEDGF Polyclonal specifically detects LEDGF in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry, Knockout Validated.
Specifications
| LEDGF | |
| Polyclonal | |
| Unconjugated | |
| O75475 | |
| PSIP1 | |
| E. coli-derived recombinant human LEDGF Met1-Asn530 Accession # O75475 | |
| 100 μg | |
| Primary | |
| Detects human LEDGF in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, Immunoassay | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunocytochemistry 5-15 ug/mL, Knockout Validated | |
| CLL-associated antigen KW-7, Dense fine speckles 70 kDa protein, DFS 70, DFS70, LEDGFDFS 70, Lens epithelium-derived growth factor, MGC74712, p52, p75, PAIP, PC4 and SFRS1 interacting protein 1, PC4 and SFRS1 interacting protein 2, PC4 and SFRS1-interacting protein, PSIP1, PSIP2, transcriptional coactivator p52/p75, Transcriptional coactivator p75/p52 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 11168 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction