missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LECT2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF722
This item is not returnable.
View return policy
Description
LECT2 Polyclonal specifically detects LECT2 in Human samples. It is validated for Western Blot.
Specifications
| LECT2 | |
| Polyclonal | |
| Unconjugated | |
| O14960 | |
| LECT2 | |
| E. coli-derived recombinant human LECT2 Gly19-Leu151 Accession # O14960 | |
| 100 μg | |
| Primary | |
| Detects human LECT2 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| chm2, chm-II, Chondromodulin II, chondromodulin-II, hLECT2, LECT-2, leukocyte cell-derived chemotaxin 2, leukocyte cell-derived chemotaxin-2, MGC126628, MIM1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3950 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction