missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LECT2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB722-SP
This item is not returnable.
View return policy
Description
LECT2 Monoclonal specifically detects LECT2 in Human samples. It is validated for Western Blot.
Specifications
| LECT2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| chm2, chm-II, Chondromodulin II, chondromodulin-II, hLECT2, LECT-2, leukocyte cell-derived chemotaxin 2, leukocyte cell-derived chemotaxin-2, MGC126628, MIM1 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3950 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 102717 | |
| Western Blot 1 ug/mL | |
| O14960 | |
| LECT2 | |
| E. coli-derived recombinant human LECT2 Gly19-Leu151 Accession # O14960 | |
| 25 μg | |
| Primary | |
| Detects recombinant human LECT2 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction