missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human LEC (CCL16) Recombinant Protein, PeproTech®

Product Code. 17892643
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 17892643

Brand: Gibco™ 30044250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human LEC is an 11.2 kDa protein containing 97 amino acid residues, including the four conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CCL16 gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. CCL16 displays chemotactic activity for lymphocytes and monocytes but not for neutrophils. This cytokine also shows a potent myelosuppressive activity and suppresses proliferation of myeloid progenitor cells. The expression of this gene is upregulated by IL-10.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O15467
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 6360
Molecular Weight (g/mol) 11.2 kDa
Name Human LEC (CCL16)
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C-C motif chemokine 16; C-C motif chemokine ligand 16; CCL16; chemokine (C-C motif) ligand 16; Chemokine CC-4; Chemokine LEC; CKb12; HCC-4; IL-10-inducible chemokine; ILINCK; LCC-1; LEC; liver CC chemokine-1; liver-expressed chemokine; LMC; Lymphocyte and monocyte chemoattractant; MGC117051; monotactin-1; MTN-1; NCC4; NCC-4; new CC chemokine 4; SCYA16; SCYL4; small inducible cytokine subfamily A (Cys-Cys), member 16; small-inducible cytokine A16
Common Name CCL16
Gene Symbol CCL16
Biological Activity Determined by its ability to chemoattract total human monocytes using a concentration range of 10-100 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence QPKVPEWVNT PSTCCLKYYE KVLPRRLVVG YRKALNCHLP AIIFVTKRNR EVCTNPNDDW VQEYIKDPNL PLLPTRNLST VKIITAKNGQ PQLLNSQ
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.