missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LDOC1 (aa 93-138) Control Fragment Recombinant Protein

Product Code. 30210984
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210984

Brand: Invitrogen™ RP107103

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (72%), Rat (72%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66429 (PA5-66429. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

LDOC1 contains a leucine zipper-like motif and a proline-rich region that shares marked similarity with an SH3-binding domain. The protein localizes to the nucleus and is down-regulated in some cancer cell lines. It is thought to regulate the transcriptional response mediated by the nuclear factor kappa B (NF-kappaB). The gene has been proposed as a tumor suppressor gene whose protein product may have an important role in the development and/or progression of some cancers.The protein encoded by this gene contains a leucine zipper-like motif and a proline-rich region that shares marked similarity with an SH3-binding domain. The protein localizes to the nucleus and is down-regulated in some cancer cell lines. It is thought to regulate the transcriptional response mediated by the nuclear factor kappa B (NF-kappaB). The gene has been proposed as a tumor suppressor gene whose protein product may have an important role in the development and/or progression of some cancers.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95751
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23641
Name Human LDOC1 (aa 93-138) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 7-Mar; BCUR1; breast cancer, up-regulated 1; Gm366; Ldoc1; LDOC1 regulator of NFKB signaling; LDOC1, regulator of NFKB signaling; Leucine zipper; leucine zipper downregulated in cancer; leucine zipper down-regulated in cancer 1; leucine zipper protein down-regulated in cancer cells; leucine zipper, down-regulated in cancer 1; Mar7; Mart7; Protein LDOC1; RGD1565644
Common Name LDOC1
Gene Symbol Ldoc1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ISLLTGEAEEWVVPYIEMDSPILGDYRAFLDEMKQCFGWDDDEDDD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.