missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LDB3 (aa 198-272) Control Fragment Recombinant Protein

Product Code. 30209756
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209756

Brand: Invitrogen™ RP100928

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83935 (PA5-83935. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a PDZ domain-containing protein. PDZ motifs are modular protein-protein interaction domains consisting of 80-120 amino acid residues. PDZ domain-containing proteins interact with each other in cytoskeletal assembly or with other proteins involved in targeting and clustering of membrane proteins. The protein encoded by this gene interacts with alpha-actinin-2 through its N-terminal PDZ domain and with protein kinase C via its C-terminal LIM domains. The LIM domain is a cysteine-rich motif defined by 50-60 amino acids containing two zinc-binding modules. This protein also interacts with all three members of the myozenin family. Mutations in this gene have been associated with myofibrillar myopathy and dilated cardiomyopathy. Alternatively spliced transcript variants encoding different isoforms have been identified; all isoforms have N-terminal PDZ domains while only longer isoforms (1, 2 and 5) have C-terminal LIM domains.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75112
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 11155
Name Human LDB3 (aa 198-272) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AW742271; cardiomyopathy, dilated 1 C (autosomal dominant); CMD1C; CMH24; CMPD3; cypher; FLJ35865; HGNC:15710; KIAA01613; KIAA0613; LDB3; LDB3Z1; LDB3Z4; LIM domain binding 3; LIM domain-binding protein 3; LVNC3; MFM4; ORACLE; oracle protein; PDLIM6; PDZ and LIM domain 6; PDZ-LIM domain protein; protein cypher; Protein oracle; RGD1564875; ZASP; Z-band alternatively spliced PDZ motif protein; Z-band alternatively spliced PDZ-motif protein
Common Name LDB3
Gene Symbol LDB3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PIGLYSAETLREMAQMYQMSLRGKASGVGLPGGSLPIKDLAVDSASPVYQAVIKSQNKPEDEADEWARRSSNLQS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.