missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lck Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB37041
This item is not returnable.
View return policy
Description
Lck Monoclonal specifically detects Lck in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| Lck | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Simple Western 2 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| P06239 | |
| LCK | |
| E. coli-derived recombinant human Lck Met1-Val66 Accession # P06239 | |
| 100 μg | |
| Primary | |
| Detects human Lck in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant rat Lck is observed. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 693010 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.10, EC 2.7.10.2, Hck-3, Lcktkr, Leukocyte C-terminal Src kinase, LSK, LSTRA, Lymphocyte cell-specific protein-tyrosine kinase, lymphocyte-specific protein tyrosine kinase, p56(LSTRA) protein-tyrosine kinase, p56lck, p56-LCK, pp58lck, Protein YT16, Proto-oncogene Lck, proto-oncogene tyrosine-protein kinase LCK, T cell-specific protein-tyrosine kinase, T-lymphocyte specific protein tyrosine kinase p56lck, tyrosine-protein kinase Lck, YT16 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3932 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction