missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lck Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3704-SP
This item is not returnable.
View return policy
Description
Lck Polyclonal specifically detects Lck in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Lck | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.10, EC 2.7.10.2, Hck-3, Lcktkr, Leukocyte C-terminal Src kinase, LSK, LSTRA, Lymphocyte cell-specific protein-tyrosine kinase, lymphocyte-specific protein tyrosine kinase, p56(LSTRA) protein-tyrosine kinase, p56lck, p56-LCK, pp58lck, Protein YT16, Proto-oncogene Lck, proto-oncogene tyrosine-protein kinase LCK, T cell-specific protein-tyrosine kinase, T-lymphocyte specific protein tyrosine kinase p56lck, tyrosine-protein kinase Lck, YT16 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3932 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 3-15 ug/mL | |
| P06239 | |
| LCK | |
| E. coli-derived recombinant human Lck Met1-Val66 Accession # P06239 | |
| 25 μg | |
| Primary | |
| Detects human Lck in Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction