missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Latent TGF-beta bp2/LTBP-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £377.00
Specifications
| Antigen | Latent TGF-beta bp2/LTBP-2 |
|---|---|
| Clone | 327318 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18754068
![]() |
R&D Systems
MAB3850-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18129820
![]() |
R&D Systems
MAB3850 |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Latent TGF-beta bp2/LTBP-2 Monoclonal specifically detects Latent TGF-beta bp2/LTBP-2 in Human samples. It is validated for Western Blot.Specifications
| Latent TGF-beta bp2/LTBP-2 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_000419 | |
| 4053 | |
| Mouse myeloma cell line NS0-derived recombinant human Latent TGF-beta bp2/LTBP-2 Gln36-Glu1821 Accession # NP_000419 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Latent TGF-beta bp2/LTBP-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse LTMP-4 is observed. |
| 327318 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C14orf141, GLC3D, Latent TGFbeta bp2, latent transforming growth factor beta binding protein 2, latent-transforming growth factor beta-binding protein 2, LTBP2, LTBP-2, LTBP3chromosome 14 open reading frame 141, MSTP031 | |
| LTBP2 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title