missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Latent TGF-beta bp2/LTBP-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3850-SP
This item is not returnable.
View return policy
Description
Latent TGF-beta bp2/LTBP-2 Polyclonal specifically detects Latent TGF-beta bp2/LTBP-2 in Human samples. It is validated for Western Blot.
Specifications
| Latent TGF-beta bp2/LTBP-2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C14orf141, GLC3D, Latent TGFbeta bp2, latent transforming growth factor beta binding protein 2, latent-transforming growth factor beta-binding protein 2, LTBP2, LTBP-2, LTBP3chromosome 14 open reading frame 141, MSTP031 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4053 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q14767 | |
| LTBP2 | |
| Mouse myeloma cell line NS0-derived recombinant human Latent TGF-beta bp2/LTBP-2 Gln36-Glu1821 Accession # Q14767 | |
| 25 μg | |
| Primary | |
| Detects human Latent TGF-beta bp2/LTBP-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse (rm) LTBP-2 is observed, and less than 1% cross-reactivity with recombinant human LTBP-1 and rmLTBP-4 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction