missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LAPTM4A (aa 187-233) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111635 (PA5-111635. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May function in the transport of nucleosides and/or nucleoside derivatives between the cytosol and the lumen of an intracellular membrane-bound compartment.
Specifications
Specifications
| Accession Number | Q15012 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 9741 |
| Name | Human LAPTM4A (aa 187-233) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | AA286466; golgi 4-transmembrane spanning transporter; golgi 4-transmembrane-spanning transporter; golgi 4-transmembrane-spanning transporter MTP; HUMORF13; KIAA0108; LAPTM4; LAPTM4A; lysosomal protein transmembrane 4 alpha; lysosomal-associated protein transmembrane 4 alpha; lysosomal-associated protein transmembrane 4 A; lysosomal-associated transmembrane protein 4 A; lysosomal-associated transmembrane protein 4-like protein; MBNT; membrane nucleoside transporter; membrane transporter protein; mKIAA0108; MTP; Mtrp; similar to lysosomal-associated protein transmembrane 4 alpha; transmembrane glycoprotein; transporter protein; UNQ1846/PRO3574; wu:fb57c07; wu:fj99e08 |
| Common Name | LAPTM4A |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction