missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LAMTOR4 (aa 1-99) Control Fragment Recombinant Protein

Product Code. 30203516
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30203516

Brand: Invitrogen™ RP92832

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54301 (PA5-54301. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

LAMTOR4 is a part of the Ragulator complex it is involved in amino acid sensing and activation of mTORC1, a signaling complex promoting cell growth in response to growth factors, energy levels, and amino acids. LAMTOR4 is activated by amino acids through a mechanism involving the lysosomal V-ATPase, the Ragulator functions as a guanine nucleotide exchange factor activating the small GTPases Rag. LAMTOR4 is activated Ragulator and Rag GTPases function as a scaffold recruiting mTORC1 to lysosomes where it is in turn activated.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q0VGL1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 389541
Name Human LAMTOR4 (aa 1-99) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 0910001L09Rik; AV006840; C7orf59; Lamtor4; Late endosomal/lysosomal adaptor and MAPK and MTOR activator 4; late endosomal/lysosomal adaptor, MAPK and MTOR activator 4; ragulator complex protein LAMTOR4; Ragulator complex protein LAMTOR4, N-terminally processed; RGD1309735; UPF0539 protein C7orf59; UPF0539 protein C7orf59 homolog
Common Name LAMTOR4
Gene Symbol LAMTOR4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MTSALTQGLERIPDQLGYLVLSEGAVLASSGDLENDEQAASAISELVSTACGFRLHRGMNVPFKRLSVVFGEHTLLVTVSGQRVFVVKRQNRGREPIDV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.