missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human LAMP-2A/CD107b (NP_002285) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Dimensione della confezione:
10µg
Descrizione
The Human LAMP-2A/CD107b (NP_002285) VersaClone cDNA clone contains the complete ORF for LAMP-2/CD107b, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The LAMP-2/CD107b insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifica
Specifica
| Gene Alias | CD107 antigen-like family member B, CD107b, CD107b antigen, LAMP2, LAMP-2, LAMPB, LGP110, LGP-96, Lysosomal Associated Membrane Protein 2, lysosomal-associated membrane protein 2, lysosome-associated membrane glycoprotein 2, Lysosome-associated membrane protein 2 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |
For Reasearch Use Only; Not for Use in Humans
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto