Learn More
Invitrogen™ Human Laminin alpha 4 ELISA Kit
Human Laminin alpha 4 quantitates human Laminin alpha 4 in serum, plasma, supernatant.
Brand: Invitrogen™ EH293RBX5
Additional Details : Weight : 0.33800kg
Description
Human Laminin alpha 4 quantitates human Laminin alpha 4 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Laminin alpha 4.
Laminin is a non-collagenous protein of basement membranes. By rotary shadowing it has a cross-like shape, consisting of one long arm, (400 kD) and three short arms (200 kD). Antibodies to laminin localize exclusively in basement membranes. Laminin is found throughout the basement membrane, but the reaction with the antibodies to laminin is more intense in the lamina lucida, the part of the basement membrane adjacent to the cell membrane. Laminin binds to type IV collagen, a component exclusively localized in the basement membrane. Laminin has been shown to play a role in cell adhesion and attachment in vivo and in vitro.Specifications
CMD1JJ; LAMA3; LAMA4; LAMA4*-1; laminin [a]4; laminin alpha 4 chain; laminin subunit alpha 4; laminin subunit alpha-4; laminin, alpha 4; laminin-14 subunit alpha; Laminin-8 subunit alpha; Laminin-9 subunit alpha; RGD1560062 | |
1.63 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
LAMA4 | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
1.63 to 400 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q16363 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
5 x 96 Tests | |
Supernatant, 50 μL; Serum, 50 μL; Plasma, 50 μL | |
Laminin alpha-4 | |
4 hr 45 min |