missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Laminin alpha 1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4187-SP
This item is not returnable.
View return policy
Description
Laminin alpha 1 Monoclonal specifically detects Laminin alpha 1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| Laminin alpha 1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| LAMA, LAMA1, LAMA1 laminin, alpha 1, S-LAM-alpha | |
| E. coli-derived recombinant human Laminin alpha 1, Leu22-Met269, Accession # P25391 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 775016 | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| P25391 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 284217 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction