missing translation for 'onlineSavingsMsg'
Learn More

Human Lamin B1 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£184.00 - £413.00

Specifications

Antigen Lamin B1
Clone 919007
Dilution Western Blot 2 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 8-25 μg/mL
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18764697
GREENER_CHOICE
View Documents
R&D Systems
MAB8525-SP
25 μg
£184.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18724627
GREENER_CHOICE
View Documents
R&D Systems
MAB8525-100
100 μg
£413.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lamin B1 Monoclonal specifically detects Lamin B1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications

Specifications

Lamin B1
Western Blot 2 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 8-25 μg/mL
Monoclonal
Supernatant
Human
P20700
4001
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
919007
Western Blot, Immunohistochemistry, Immunocytochemistry
Unconjugated
Mouse
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
ADLD, lamin B1, lamin-B1, LMN, LMN2, LMNB, LMNB1, MGC111419
E. coli-derived recombinant human Lamin B1 Arg418-Ser582 Accession # P20700
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Lamin B1 in direct ELISAs.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.