missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Lamin B Receptor Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (62%), Rat (62%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66473 (PA5-66473. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The protein encoded by this gene belongs to the ERG4/ERG24 family. It localized in the nuclear envelope inner membrane and anchors the lamina and the heterochromatin to the membrane. It may mediate interaction between chromatin and lamin B. Mutations of this gene has been associated with autosomal recessive HEM/Greenberg skeletal dysplasia. Alternative splicing occurs at this locus and two transcript variants encoding the same protein have been identified.
Specifications
Specifications
| Accession Number | Q14739 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 3930 |
| Name | Human Lamin B Receptor Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 3-beta-hydroxysterol Delta (14)-reductase; AI505894; C-14 sterol reductase; C14SR; Delta(14)-sterol reductase LBR; delta(14)-sterol reductase LBR; lamin-B receptor; Delta-14-SR; DHCR14B; FLJ43126; ic; ichthyosis; integral nuclear envelope inner membrane protein; lamin B receptor; lamin-B receptor; lamin-b receptor-like protein; Lbr; LMN2R; MGC9041; NBP60; PHA; PRO0650; sb:cb406; Sterol C14-reductase; TDRD18; tudor domain containing 18; wu:fb75h08; wu:fc47b04; wu:fd36b07; zgc:86649 |
| Common Name | Lamin B Receptor |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction