missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LAIR2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2665
This item is not returnable.
View return policy
Description
LAIR2 Polyclonal specifically detects LAIR2 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| LAIR2 | |
| Polyclonal | |
| Unconjugated | |
| Q6ISS4 | |
| LAIR2 | |
| Mouse myeloma cell line NS0-derived recombinant human LAIR2 Gln22-Pro152 Accession # Q6ISS4 | |
| 100 μg | |
| Primary | |
| Detects human LAIR2 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human LAIR1 is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| CD306, CD306 antigen, CD306leukocyte-associated Ig-like receptor-2, LAIR-2, leukocyte-associated Ig-like receptor 2, leukocyte-associated immunoglobulin-like receptor 2, MGC71634 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3904 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction