missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LAIR1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2664-SP
This item is not returnable.
View return policy
Description
LAIR1 Polyclonal specifically detects LAIR1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| LAIR1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD305, CD305 antigen, CD305leukocyte-associated Ig-like receptor 1, hLAIR1, LAIR-1, leukocyte-associated immunoglobulin-like receptor 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3903 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q6GTX8 | |
| LAIR1 | |
| Mouse myeloma cell line NS0-derived recombinant human LAIR1 isoform 1 Gln22-His163 Accession # Q6GTX8 | |
| 25 μg | |
| Primary | |
| Detects human LAIR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant human LAIR2 is observed and less than 2% cross-reactivity with recombinant mouse LAIR1 is observed. | |
| Human | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur