missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LAG-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB23196-100
This item is not returnable.
View return policy
Description
LAG-3 Monoclonal specifically detects LAG-3 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| LAG-3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| CD223, CD223 antigen, FDC, LAG3, LAG-3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Protein FDC | |
| Mouse myeloma cell line NS0-derived recombinant human LAG-3, Leu23-Leu450, Accession # P18627 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 874512 | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, CyTOF-ready | |
| P18627 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3902 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction