missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human L1TD1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB8317
This item is not returnable.
View return policy
Description
L1TD1 Monoclonal specifically detects L1TD1 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| L1TD1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q5T7N2 | |
| L1TD1 | |
| E. coli-derived recombinant human L1TD1 Met1-Leu142 Accession # Q5T7N2 | |
| 100 μg | |
| Primary | |
| Detects human L1TD1 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 901258 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ECAT11, ES cell-associated protein 11, FLJ10884, LINE-1 type transposase domain containing 1, LINE-1 type transposase domain-containing protein 1, MGC133253, RP5-1155K23.3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 54596 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction