missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human L-Myc (aa 33-88) Control Fragment Recombinant Protein

Product Code. 30211629
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30211629

Brand: Invitrogen™ RP105255

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66558 (PA5-66558. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Cellular DNA-binding proteins encoded by the c-myc genes. They are normally involved in nucleic acid metabolism and in mediating the cellular response to growth factors. Elevated and deregulated (constitutive) expression of c-myc proteins can cause tumorigenesis. L-Myc gene ontology annotations include DNA binding function, DNA-binding transcription factor activity, RNA polymerase II-specific action, protein dimerization activity, and regulation of inner ear auditory receptor cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P12524
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 4610
Name Human L-Myc (aa 33-88) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Avian myelocytomatosis viral (v-myc) related oncogene homolog 1, lung carcinoma derived (Lmyc); AW536278; bHLHe38; Class E basic helix-loop-helix protein 38; LMYC; L-MYC; Lmyc1; Lmyc-1; l-myc-1 proto-oncogene; MYCL; MYCL proto-oncogene, bHLH transcription factor; Mycl1; myc-related gene from lung cancer; Protein L-Myc; protein L-Myc-1; v-myc avian myelocytomatosis viral oncogene lung carcinoma derived homolog; V-myc myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog 1, lung carcinoma derived; v-myc myelocytomatosis viral oncogene homolog, lung carcinoma derived (avian)
Common Name L-Myc
Gene Symbol MYCL
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FELVPSPPTSPPWGLGPGAGDPAPGIGPPEPWPGGCTGDEAESRGHSKGWGRNYAS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.