missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human KRT82 (aa 62-112) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84752 (PA5-84752. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
KRT82 is a member of the keratin gene family. As a type II hair keratin, it is a basic protein which heterodimerizes with type I keratins to form hair and nails. The type II hair keratins are clustered in a region of chromosome 12q13 and are grouped into two distinct subfamilies based on structure similarity. One subfamily, consisting of KRTHB1, KRTHB3, and KRTHB6, is highly related. The other less-related subfamily includes KRTHB2, KRTHB4, and KRTHB5. All hair keratins are expressed in the hair follicle; this keratin appears to be a hair cuticle-specific keratin.
Specifications
Specifications
| Accession Number | Q9NSB4 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 3888 |
| Name | Human KRT82 (aa 62-112) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | hard keratin, type II, 2; HB2; Hb-2; K82; Kb22; keratin 82; keratin 82, type II; keratin complex 2, basic, gene 20; keratin, hair, basic, 2; keratin, type II cuticular Hb2; Keratin-82; Krt2-20; Krt82; Krthb2; Type II hair keratin Hb2; type II keratin Kb22; type-II keratin Kb22 |
| Common Name | KRT82 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction