missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human KRT81 (aa 293-326) Control Fragment Recombinant Protein

Product Code. 30205821
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205821

Brand: Invitrogen™ RP105621

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65582 (PA5-65582. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a member of the keratin gene family. As a type II hair keratin, it is a basic protein which heterodimerizes with type I keratins to form hair and nails. The type II hair keratins are clustered in a region of chromosome 12q13 and are grouped into two distinct subfamilies based on structure similarity. One subfamily, consisting of KRTHB1, KRTHB3, and KRTHB6, is highly related. The other less-related subfamily includes KRTHB2, KRTHB4, and KRTHB5. All hair keratins are expressed in the hair follicle; this hair keratin, as well as KRTHB3 and KRTHB6, is found primarily in the hair cortex. Mutations in this gene and KRTHB6 have been observed in patients with a rare dominant hair disease, monilethrix.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14533
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3887
Name Human KRT81 (aa 293-326) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA589625; ghHb1; ghHkb1; Hair keratin K2.9; hard keratin, type II, 1; HB1; Hb-1; hHAKB2-1; K81; Kb21; keratin 81; keratin 81, type II; keratin complex 2, basic, gene 19; keratin, hair, basic, 1; Keratin, type II cuticular Hb1; keratin-81; Krt2-19; KRT81; KRTHB1; Metastatic lymph node 137 gene protein; MLN 137; MLN137; type II hair keratin Hb1; type II keratin Kb21; type-II keratin Kb21
Common Name KRT81
Gene Symbol KRT81
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence AESWYRSKCEEMKATVIRHGETLRRTKEEINELN
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.