missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kremen-2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF1946
This item is not returnable.
View return policy
Description
Kremen-2 Polyclonal specifically detects Kremen-2 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| Kremen-2 | |
| Polyclonal | |
| Unconjugated | |
| Q8NCW0 | |
| KREMEN2 | |
| Mouse myeloma cell line NS0-derived recombinant human Kremen-2 Gln19-Ala364 Accession # Q8NCW0 | |
| 100 μg | |
| Primary | |
| Detects human Kremen-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 35% cross-reactivity with recombinant mouse Kremen-2 is observed and 25% cross-reactivity with recombinant human Kremen-1 is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Dickkopf receptor 2, kremen protein 2, Kremen2, kringle containing transmembrane protein 2, Kringle domain-containing transmembrane protein 2, KRM2Kringle-containing protein marking the eye and the nose, MGC10791, MGC16709 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 79412 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction