missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human KPNA3 Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
KPNA3 Synthetic Peptide, PEP-1149, from Invitrogen.
The transport of molecules between the nucleus and the cytoplasm in eukaryotic cells is mediated by the nuclear pore complex (NPC), which consists of 60-100 proteins. Small molecules (up to 70 kD) can pass through the nuclear pore by nonselective diffusion while larger molecules are transported by an active process. The protein encoded by this gene belongs to the importin alpha family, and is involved in nuclear protein import.
Specifications
Specifications
| Gene ID (Entrez) | 3839 |
| Species | Human |
| Purification Method | purified |
| Accession Number | O00505 |
| Common Name | Human KPNA3 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction