missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KOR Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB38951-SP
This item is not returnable.
View return policy
Description
KOR Monoclonal specifically detects KOR in Human samples. It is validated for Immunohistochemistry.
Specifications
| KOR | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P41145 | |
| OPRK1 | |
| NS0 mouse myeloma cell line transfected with human KOR Met1-Val380 Accession # P41145 | |
| 25 μg | |
| Primary | |
| Detects human KOR. Stains human KOR transfectants but not irrelevant transfectants. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 387310 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| kappa opioid receptor, kappa-type opioid receptor, KOR, K-OR-1, KOR-1, Opiate receptor, kappa-1, opioid receptor, kappa 1, OPRK, OPRK1, Oprk2, R21 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4986 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction