missing translation for 'onlineSavingsMsg'
Learn More

Human KLRG1 Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

£181.00 - £444.00

Specifications

Antigen KLRG1
Clone 2388C
Dilution Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 3-25 μg/mL, CyTOF-ready
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18731577
GREENER_CHOICE
View Documents
R&D Systems
MAB70293-SP
25 μg
£181.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18113381
GREENER_CHOICE
View Documents
R&D Systems
MAB70293-100
100 μg
£444.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

KLRG1 Monoclonal specifically detects KLRG1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications

Specifications

KLRG1
Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 3-25 μg/mL, CyTOF-ready
Monoclonal
Supernatant
Human
Q96E93
10219.0
IgG
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
2388C
Flow Cytometry, Immunohistochemistry, CyTOF
Unconjugated
Rabbit
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
2F1, CLEC15A, CLEC15AMAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, killer cell lectin-like receptor subfamily G member 1, killer cell lectin-like receptor subfamily G, member 1, MAFA, MAFAL, MAFA-L, MAFA-like receptor, MAFAMAFA-2F1, Mast cell function-associated antigen, mast cell function-associated antigen (ITIM-containing), MGC13600
Chinese hamster ovary cell line CHO-derived recombinant human KLRG1, Leu60-Phe195, Accession No. Q96E93
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human KLRG1 in direct ELISAs.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.