missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Klotho beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5889
This item is not returnable.
View return policy
Description
Klotho beta Monoclonal specifically detects Klotho beta in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| Klotho beta | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta-klotho, betaKlotho, BKL, KLB, klotho beta, klotho beta like, klotho beta-like protein, MGC142213 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 152831 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 885935 | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| Q86Z14 | |
| KLB | |
| Mouse myeloma cell line NS0-derived recombinant human Klotho beta Phe53-Leu997 Accession # Q86Z14 | |
| 100 μg | |
| Primary | |
| Detects human Klotho beta in direct ELISA and Western blots. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction