missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KLF12 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems IC9097U-100UG
This item is not returnable.
View return policy
Description
KLF12 Monoclonal specifically detects KLF12 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| KLF12 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| AP-2rep, AP2REP, AP-2rep transcription factor, AP2REPAP-2 repressor, HSPC122, KLF12 zinc finger transcriptional repressor, Krueppel-like factor 12, Kruppel-like factor 12, Transcriptional repressor AP-2rep | |
| Mouse myeloma cell line NS0-derived human KLF12 Asn2-Val402 Accession # Q9Y4X4 | |
| RUO | |
| 11278 | |
| Human | |
| IgG |
| Flow Cytometry | |
| 1230C | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9Y4X4 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Detects human KLF12 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction