missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kirrel2/NEPH3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF2564
This item is not returnable.
View return policy
Description
Kirrel2/NEPH3 Polyclonal specifically detects Kirrel2/NEPH3 in Human samples. It is validated for Western Blot.
Specifications
| Kirrel2/NEPH3 | |
| Polyclonal | |
| Unconjugated | |
| Q6UWL6 | |
| KIRREL2 | |
| Mouse myeloma cell line NS0-derived recombinant human Kirrel2/NEPH3 Gly21-Arg503 Accession # Q6UWL6 | |
| 100 μg | |
| Primary | |
| Detects human Kirrel2/NEPH3 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse Kirrel2 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| DKFZp564A1164, Filtrin, FILTRINkin of IRRE-like protein 2, kin of IRRE like 2 (Drosophila), Kin of irregular chiasm-like protein 2, MGC15718, NEPH3, NEPH3Nephrin-like protein 3, nephrin-like 3, nephrin-like gene 1, NLG1, NLG1kin of irregular chiasm-like 2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 84063 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction