missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Kir3.1 (KCNJ3) (aa 351-487) Control Fragment Recombinant Protein

Product Code. 30212301
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30212301

Brand: Invitrogen™ RP90677

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This potassium channel is controlled by G proteins. Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. This receptor plays a crucial role in regulating the heartbeat.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P48549
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3760
Name Human Kir3.1 (KCNJ3) (aa 351-487) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias EGK_04447; G protein-activated inward rectifier potassium channel 1; GIRK1; GIRK-1; HGIRK1; inward rectifier K(+) channel Kir3.1; inward rectifier K+ channel KIR3.1; Kcnf3; Kcnj3; KGA; KGB1; KIR3.1; potassium channel inwarding rectifying channel subfamily J member 3; potassium channel subunit Kir3.1 type 3 delta; potassium channel, inwardly rectifying subfamily J member 3; potassium channel, inwardly rectifying subfamily J, member 3; potassium inwardly rectifying channel subfamily J member 3; potassium inwardly-rectifying channel subfamily J member 3 splice variant 1 e; potassium inwardly-rectifying channel, subfamily J, member 3; potassium voltage-gated channel subfamily J member 3
Common Name Kir3.1 (KCNJ3)
Gene Symbol KCNJ3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VPTPPYSVKEQEEMLLMSSPLIAPAITNSKERHNSVECLDGLDDITTKLPSKLQKITGREDFPKKLLRMSSTTSEKAYSLGDLPMKLQRISSVPGNSEEKLVSKTTKMLSDPMSQSVADLPPKLQKMAGGAARMEGN
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.