missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KIR2DS4/CD158i Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£168.00 - £550.00
Specifications
| Antigen | KIR2DS4/CD158i |
|---|---|
| Clone | 179315 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18720918
![]() |
R&D Systems
MAB1847-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18172261
![]() |
R&D Systems
MAB1847 |
500 μg |
£550.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KIR2DS4/CD158i Monoclonal specifically detects KIR2DS4/CD158i in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| KIR2DS4/CD158i | |
| LYOPH | |
| Western Blot, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| P43632 | |
| 3809 | |
| BaF3 mouse pro-B cell line transfected with human KIR2DS4 Accession # P43632 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects KIR2DS4/CD158i in Western blots. In flow cytometry, this antibody stains cells transfected with KIR2DS4/CD158i. Does not stain cells transfected with KIR2DL1, 2DL2, 2DL3, 2DL4, 2DL5, 2DS1, 2DS2, 3DL1, 3DL2, or 3DS1. |
| 179315 | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD158 antigen-like family member I, CD158I, CD158i antigen, cl-39, killer cell immunoglobulin-like receptor 2DS4, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail4,nkat8, killer inhibitory receptor 4-1-2, KIR antigen 2DS4, KKA3KIR1D, MGC120019, MGC125315, MGC125317, MHC class I NK cell receptor, natural killer cell inhibitory receptor, Natural killer-associated transcript 8, NKAT-8, NKAT8KIR412, P58 natural killer cell receptor clones CL-39/CL-17, p58 NK receptor CL-39/CL-17 | |
| KIR2DS4 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title