missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KIR2DS1/CD158h Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB8887-SP
This item is not returnable.
View return policy
Description
KIR2DS1/CD158h Monoclonal specifically detects KIR2DS1/CD158h in Human samples. It is validated for Flow Cytometry.
Specifications
| KIR2DS1/CD158h | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| killer cell immunoglobulin-like receptor 2DS1, CD158a, CD158H, EB6ActI, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 1, p50.1 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3806 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Flow Cytometry | |
| 1127B | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Q14954 | |
| KIR2DS1 | |
| HEK293 human embryonic kidney cell line transfected with human KIR2DS1/CD158h Met1-His245 Accession # Q14954 | |
| 25 μg | |
| Primary | |
| Detects human KIR2DS1/CD158h in flow cytometry. Clone 1127B recognizes KIR2DS1 and some alleles of KIR2DL1, both members of the killer cell immunoglobulin-like receptor (KIR) family. Because Clone 1127B displays partial cross-reactivity with KIR2DL1, co-staining with Mouse Anti-Human KIR2DL1 Clone 143211 (Catalog # FAB1844F) is recommended. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction