Learn More
Human Kininogen/Kininostatin Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 3 publications
Brand:  R&D Systems AF1396-SP
 View more versions of this product
Product Code. 18717177
  • £168.00 / 25µg
Description
Kininogen/Kininostatin/Bradykinin Polyclonal specifically detects Kininogen/Kininostatin/Bradykinin in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Kininogen/Kininostatin/Bradykinin
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
alpha 2-thiol Proteinase Inhibitor, Alpha-2-thiol proteinase inhibitor, BDK, bradykinin, Fitzgerald factor, High molecular weight kininogen, kininogen, kininogen 1, kininogen-1, KNG1, KNGHMWK, Williams-Fitzgerald-Flaujeac factor
Goat
Affinity Purified
RUO
3827
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot
LYOPH
Western Blot 0.1 ug/mL
P01042
KNG1
E. coli-derived recombinant human Kininostatin Lys438-Ser531 Accession # P01042
25 μg
Primary
Detects human Kininogen/Kininostatin in direct ELISAs and Western blots.