missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kininogen/Kininostatin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF1396
This item is not returnable.
View return policy
Description
Kininogen/Kininostatin/Bradykinin Polyclonal specifically detects Kininogen/Kininostatin/Bradykinin in Human samples. It is validated for Western Blot.
Specifications
| Kininogen/Kininostatin/Bradykinin | |
| Polyclonal | |
| Unconjugated | |
| P01042 | |
| KNG1 | |
| E. coli-derived recombinant human Kininostatin Lys438-Ser531 Accession # P01042 | |
| 100 μg | |
| Primary | |
| Detects human Kininogen/Kininostatin in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| alpha 2-thiol Proteinase Inhibitor, Alpha-2-thiol proteinase inhibitor, BDK, bradykinin, Fitzgerald factor, High molecular weight kininogen, kininogen, kininogen 1, kininogen-1, KNG1, KNGHMWK, Williams-Fitzgerald-Flaujeac factor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3827 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction