missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kininogen Heavy Chain Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB15692
Additional Details : Weight : 0.00970kg
Description
Kininogen Monoclonal specifically detects Kininogen in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Kininogen | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
P01042 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3827 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Immunoprecipitation | |
236012 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
KNG1 | |
Mouse myeloma cell line NS0-derived recombinant human Kininogen Gln19-Ser644 (Ile581Thr) Accession # P01042 | |
500 μg | |
Primary | |
Detects human Kininogen Heavy Chain in direct ELISAs and Western blots. It reacts with HK, Kininogen, and the heavy chain, but not with the light chain alone. In direct ELISAs, no cross-reactivity with recombinant human (rh) Cystatin A, B, C, D, E, S, SA, SN, rhFetuin A, rhFetuin B, rhHPRG, recombinant mouse Kininogen, or rhKininostatin is observed. | |
Human | |
Purified |
For Research Use Only