missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Kinesin 5B (aa 376-396) Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Human Kinesin 5B (residues 376-396) Synthetic Peptide, PEP-198, from Invitrogen.
Microtubule-dependent motor required for normal distribution of mitochondria and lysosomes. Can induce formation of neurite-like membrane protrusions in non-neuronal cells in a ZFYVE27-dependent manner. Regulates centrosome and nuclear positioning during mitotic entry. During the G2 phase of the cell cycle in a BICD2-dependent manner, antagonizes dynein function and drives the separation of nuclei and centrosomes. Required for anterograde axonal transportation of MAPK8IP3/JIP3 which is essential for MAPK8IP3/JIP3 function in axon elongation.
Specifications
Specifications
| Gene ID (Entrez) | 3799 |
| Species | Human |
| Purification Method | purified |
| Accession Number | P33176 |
| Common Name | Human Kinesin 5 B (aa 376-396) |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C. Avoid freeze-thaw cycles. |
| Description | Synthetic Peptide |
| Format | Lyophilized |
| Formulation | peptide with no preservative |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction