missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Kidney Biomarker Premixed Magnetic Luminex™ Performance Assay

Product Code. 18401290
Change view
Click to view available options
Quantity:
2 Plex
3 Plex
4 Plex
5 Plex
6 Plex
7 Plex
8 Plex
Unit Size:
1 set
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401290 8 Plex 1 set
18422270 7 Plex 1 set
18492610 5 Plex 1 set
18401790 6 Plex 1 set
18461740 3 Plex 1 set
18432510 2 Plex 1 set
18482060 4 Plex 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 18401290 Supplier R&D Systems™ Supplier No. FCSTM1608

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Magnetic bead-based multiplex assay for the Luminex™ platform

TRUSTED_SUSTAINABILITY

Specifications

Quantity 8 Plex
Species Human
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.