missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KIAA1217 Full-length ORF (AAH18764.1, 1 a.a. - 877 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16102163
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16102163 10 μg 10µg
16112163 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16102163 Supplier Abnova™ Supplier No. H00056243P01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

Sequence: MVVPGNATIPRDRISSLPVSRPISPSPSAILERRDVKPDEDMSGKNIAMYRNEGFYADPYLYHEGRMSIASSHGGHPLDVPDHIIAYHRTAIRSASAYCNPSMQAEMHMEQSLYRQKSRKYPDSHLPTLGSKTPPASPHRVSDLRMIDMHAHYNAHGPPHTMQPDRASPSRQAFKKEPGTLVYIEKPRSAAGLSSLVDLGPPLMEKQVFAYSTATIPKDRETSEKMMKTTANRNHTDSAGTPHVSGGKMLSALESTVPPSQPPPVGTSAIHMSLLEMRRSVAELRLQLQQMRQLQLQNQELLRAMMKKAELEISGKVMETMKRLEDPVQRQRVLVEQERQKYLHEEEKIVKKLCELEDFVEDLKKDSTAASRLVTLKDVEDGAFLLRQVGEAVATLKGEFPTLQNKMRAILRIEVEAVRFLKEEPHKLDSLLKRVRSMTDVLTMLRRHVTDGLLKGTDAAQAAQYMAMEKATAAEVLKSQEEAAHTSGQPFHSTGAPGDAKSEVVPLSGMMVRHAQSSPVVIQPSQHSVALLNPAQNLPHVASSPAVPQEATSTLQMSQAPQSPQIPMNGSAMQSLFIEEIHSVSAKNRAVSIEKAEKKWEEKRQNLDHYNGKEFEKLLEEAQANIMKSIPNLEMPPATGPLPRGDAPVDKVELSEDSPNSEQDLEKLGGKSPPPPPPPPRRSYLPGSGLTTTRSGDVVYTGRKENITAKASSEDAGPSPQTRATKYPAEEPASAWTPSPPPVTTSSSKDEEEEEEEGDKIMAELQAFQKCSFMDVNSNSHAEPSRADSHVKDTRSGATVPPKEKKNLEFFHEDVRKSDVEYENGPQMEFQKGSSGAPQTSRMPVPMSAKNRPGTLDKPGKQSKLQDPRQYRQVVLP

Specifications

Accession Number AAH18764.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 56243
Molecular Weight (g/mol) 123.4kDa
Name KIAA1217 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MVVPGNATIPRDRISSLPVSRPISPSPSAILERRDVKPDEDMSGKNIAMYRNEGFYADPYLYHEGRMSIASSHGGHPLDVPDHIIAYHRTAIRSASAYCNPSMQAEMHMEQSLYRQKSRKYPDSHLPTLGSKTPPASPHRVSDLRMIDMHAHYNAHGPPHTMQPDRASPSRQAFKKEPGTLVYIEKPRSAAGLSSLVDLGPPLMEKQVFAYSTATIPKDRETSEKMMKTTANRNHTDSAGTPHVSGGKMLSALESTVPPSQPPPVGTSAIHMSLLEMRRSVAELRLQLQQMRQLQLQNQELLRAMMKKAELEISGKVMETMKRLEDPVQRQRVLVEQERQKYLHEEEKIVKKLCELEDFVEDLKKDSTAASRLVTLKDVEDGAFLLRQVGEAVATLKGEFPTLQNKMRAILRIEVEAVRFLKEEPHKLDSLLKRVRSMTDVLTMLRRHVTDGLLKGTDAAQAAQYMAMEKATAAEVLKSQEEAAHTSGQPFHSTGAPGDAKSEVVPLSGMMVRHAQSSPVVIQPSQHSVALLNPAQNLPHVASSPAVPQEATSTLQMSQAPQSPQIPMNGSAMQSLFIEEIHSVSAKNRAVSIEKAEKKWEEKRQNLDHYNGKEFEKLLEEAQANIMKSIPNLEMPPATGPLPRGDAPVDKVELSEDSPNSEQDLEKLGGKSPPPPPPPPRRSYLPGSGLTTTRSGDVVYTGRKENITAKASSEDAGPSPQTRATKYPAEEPASAWTPSPPPVTTSSSKDEEEEEEEGDKIMAELQAFQKCSFMDVNSNSHAEPSRADSHVKDTRSGATVPPKEKKNLEFFHEDVRKSDVEYENGPQMEFQKGSSGAPQTSRMPVPMSAKNRPGTLDKPGKQSKLQDPRQYRQVVLP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp761L0424/MGC31990/SKT
Common Name KIAA1217
Gene Symbol KIAA1217
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.