missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human KIAA0556 (aa 152-245) Control Fragment Recombinant Protein

Product Code. 30201887
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201887

Brand: Invitrogen™ RP94967

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (52%), Rat (52%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57152 (PA5-57152. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a novel, evolutionarily conserved, ciliary protein. In human hTERT-RPE1 cells, the protein is found at the base of cilia, decorating the ciliary axoneme, and enriched at the ciliary tip. The protein binds to microtubules in vitro and regulates their stability when it is overexpressed. A null mutation in this gene has been associated with Joubert syndrome, a recessive disorder that is characterized by a distinctive mid-hindbrain and cerebellar malformation and is also often associated with wider ciliopathy symptoms. Consistently, in a serum-starvation ciliogenesis assay, human fibroblast cells derived from patients with the mutation display a reduced number of ciliated cells with abnormally long cilia.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O60303
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23247
Name Human KIAA0556 (aa 152-245) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias JBTS26; Katanin-interacting protein; Katnip; KIAA0556; Protein KIAA0556
Common Name KIAA0556
Gene Symbol KIAA0556
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EPPVDYSDDFELCGDVTLQANNTSEDRPQELRRSLELSVNLQRKQKDCSSDEYDSIEEDILSEPEPEDPALVGHPRHDRPPSSGDWTQKDVHGE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.