missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human KIAA0513 (aa 134-271) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52997 (PA5-52997. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The function of this protein remains unknown.
Specifications
Specifications
| Accession Number | O60268 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 9764 |
| Name | Human KIAA0513 (aa 134-271) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 6430548M08Rik; AW049007; KIAA0513; mKIAA0513; RGD1304884; RIKEN cDNA 6430548M08 gene; similar to RIKEN cDNA 6430548M08; uncharacterized protein KIAA0513; uncharacterized protein KIAA0513 homolog |
| Common Name | KIAA0513 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction