missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human KDELC1 (aa 27-97) Control Fragment Recombinant Protein

Codice prodotto. 30195379
Click to view available options
Quantity:
100 μL
Dimensione della confezione:
100µL
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 30195379

Marca: Invitrogen™ RP106057

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65374 (PA5-65374. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Protein glucosyltransferase that catalyzes the transfer of glucose from UDP-glucose to a serine residue within the consensus sequence peptide C-X-N-T-X-G-S-F-X-C. Can also catalyze the transfer of xylose from UDP-xylose but less efficiently. Specifically targets extracellular EGF repeats of proteins such as NOTCH1 and NOTCH3. May regulate the transport of NOTCH1 and NOTCH3 to the plasma membrane and thereby the Notch signaling pathway.
TRUSTED_SUSTAINABILITY

Specifica

Accession Number Q6UW63
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 79070
Name Human KDELC1 (aa 27-97) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias endoplasmic reticulum resident protein 58; EP58; ER protein 58; ERp58; KDEL (Lys-Asp-Glu-Leu) containing 1; KDEL motif containing 1; KDEL motif-containing 1; KDEL motif-containing protein 1; KDEL1; KDELC1; POGLUT2; Protein O-glucosyltransferase 2; Protein O-xylosyltransferase POGLUT2; UNQ1910/PRO4357
Common Name KDELC1
Gene Symbol KDELC1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LSPEKSEIWGPGLKADVVLPARYFYIQAVDTSGNKFTSSPGEKVFQVKVSAPEEQFTRVGVQVLDRKDGSF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato