missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human KCTD10 (aa 242-313) Control Fragment Recombinant Protein

Product Code. 30201525
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201525

Brand: Invitrogen™ RP90727

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53138 (PA5-53138. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

KCTD10, a rat potassium channel tetramerisation domain-containing 10 gene is a novel member of the polymerase delta-interacting protein 1 (PDIP1) gene family. KCTD10 shares significant similarity in amino acid sequence to PDIP1 and can interact with the small subunit of DNA polymerase delta and PCNA as PDIP1 does. Like PDIP1, the expression of KCTD10 gene can be induced by TNF-alpha in NIH3T3 cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9H3F6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 83892
Name Human KCTD10 (aa 242-313) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AW536343; BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 3; BTB/POZ domain-containing protein KCTD10; BTBD28; C87062; hBACURD3; KCTD10; mBACURD3; MSTP028; potassium channel tetramerisation domain containing 10; potassium channel tetramerization domain containing 10; potassium channel tetramerization domain-containing 10; potassium channel tetramerization domain-containing protein 10; ULR061; ULRO61; wu:fb30g12; zgc:63846
Common Name KCTD10
Gene Symbol KCTD10
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VEFPEARIYEETLNILLYEAQDGRGPDNALLEATGGAAGRSHHLDEDEERERIERVRRIHIKRPDDRAHLHQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.