missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human KCNQ2 (aa 622-696) Control Fragment Recombinant Protein

Product Code. 30201169
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201169

Brand: Invitrogen™ RP103292

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

KCNQs are members of the voltage-dependent non-inactivating potassium channel family. Currently there are five known KNCQs (KCNQ1-5) found in the central nervous system and KNCQ2 and 3 have demonstrated their importance in M-current activation. Studies have shown that KCNQ2 and KCNQ3 form heteromultimers that, when formed, substantially increase the M-current. Inhibition of M-current controls neuron excitability throughout the nervous system as well as the responsiveness to synaptic inputs. Genetic mutations in these proteins have been linked to disorders such as benign familial neonatal convulsions (BFNC), deafness, neuropathic pain and epilepsy.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43526
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3785
Name Human KCNQ2 (aa 622-696) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias BFNC; EBN; EBN1; ENB1; HNSPC; KCNA11; KCNQ2; KQT like 2; KQT2; KQT-like 2; KV7.2; KVEBN1; mKQT2.3; mKQT2.4; neuroblastoma-specific potassium channel subunit alpha KvLQT2; Nmf134; potassium channel subunit alpha KvLQT2; potassium channel, voltage gated KQT-like subfamily Q, member 2; potassium channel, voltage-gated KQT-like subfamily Q, member 2; potassium voltage-gated channel KQT-like subfamily member 2.3; potassium voltage-gated channel KQT-like subfamily member 2.4; potassium voltage-gated channel subfamily KQT member 2; potassium voltage-gated channel subfamily Q member 2; potassium voltage-gated channel, KQT-like subfamily, member 2; potassium voltage-gated channel, subfamily Q, member 2; voltage-gated potassium channel subunit Kv7.2
Common Name KCNQ2
Gene Symbol KCNQ2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RLGKVEKQVLSMEKKLDFLVNIYMQRMGIPPTETEAYFGAKEPEPAPPYHSPEDSREHVDRHGCIVKIVRSSSST
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.