missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human KCNK17 (aa 274-332) Control Fragment Recombinant Protein

Product Code. 30195033
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195033

Brand: Invitrogen™ RP94808

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (34%), Rat (34%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60543 (PA5-60543. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene belongs to the family of potassium channel proteins containing two pore-forming P domains. This channel is an open rectifier which primarily passes outward current under physiological K+ concentrations. This gene is activated at alkaline pH. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2008].
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96T54
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 89822
Name Human KCNK17 (aa 274-332) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2 P domain potassium channel Talk-2; Acid-sensitive potassium channel protein TASK-4; K2p17.1; KCNK17; Potassium channel subfamily K member 17; potassium channel, two pore domain subfamily K, member 17; potassium two pore domain channel subfamily K member 17; TALK2; TALK-2; TASK4; TASK-4; TWIK-related acid-sensitive K(+) channel 4; TWIK-related alkaline pH-activated K(+) channel 2; UNQ5816/PRO19634
Common Name KCNK17
Gene Symbol KCNK17
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence CSCCHHSSKEDFKSQSWRQGPDREPESHSPQQGCYPEGPMGIIQHLEPSAHAAGCGKDS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.