missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KCC2/SLC12A5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8369
This item is not returnable.
View return policy
Description
KCC2/SLC12A5 Monoclonal specifically detects KCC2/SLC12A5 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| KCC2/SLC12A5 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Electroneutral potassium-chloride cotransporter 2, hKCC2, KCC2K-Cl cotransporter 2, KIAA1176erythroid K-Cl cotransporter 2, Neuronal K-Cl cotransporter, SLC12A5, solute carrier family 12 (potassium/chloride transporter), member 5, solute carrier family 12 member 5 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 57468 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Flow Cytometry, CyTOF | |
| 905705 | |
| Western Blot 0.5 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q9H2X9 | |
| SLC12A5 | |
| NS0 mouse myeloma cell line transfected with human SLC12A5 Accession # Q9H2X9 | |
| 100 μg | |
| Primary | |
| Detects human SLC12A5 in ELISAs and Western blots. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction