missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kallikrein 6/Neurosin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF2008
This item is not returnable.
View return policy
Description
Kallikrein 6/Neurosin Polyclonal specifically detects Kallikrein 6/Neurosin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| Kallikrein 6/Neurosin | |
| Polyclonal | |
| Unconjugated | |
| Q92876 | |
| KLK6 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 6 Glu17-Lys244 Accession # Q92876 | |
| 100 μg | |
| Primary | |
| Detects human Kallikrein 6 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Kallikrein 3, rhKallikrein 5, rhKallikrein 11, rhKallikrein 13, rhKallikrein 14, and rhKallikrein 15 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
| Bssp, EC 3.4.21, EC 3.4.21.-, hK6, kallikrein 6 (neurosin, zyme), kallikrein-6, kallikrein-related peptidase 6, KLK6, Klk7, neurosin, Protease M, protease, serine, 18, PRSS18, PRSS9, PRSS9MGC9355, Serine protease 18, Serine protease 9, SP59, Zyme | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5653 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction