missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kallikrein 6/Neurosin Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB2008
Additional Details : Weight : 0.00970kg
Description
Kallikrein 6/Neurosin Monoclonal specifically detects Kallikrein 6/Neurosin in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Kallikrein 6/Neurosin | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Q92876 | |
KLK6 | |
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 6/Neurosin Leu22-Lys244 Accession # Q92876 | |
100 μg | |
Primary | |
Detects human Kallikrein 6/Neurosin in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Factor X, recombinant mouse HGFA, rhKLK3, 4, 5, 8, 10, 11, 15, rhThrombin, or rhuPA is observed. | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
265508 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Bssp, EC 3.4.21, EC 3.4.21.-, hK6, kallikrein 6 (neurosin, zyme), kallikrein-6, kallikrein-related peptidase 6, KLK6, Klk7, neurosin, Protease M, protease, serine, 18, PRSS18, PRSS9, PRSS9MGC9355, Serine protease 18, Serine protease 9, SP59, Zyme | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5653 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
For Research Use Only