missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kallikrein 5 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1108-SP
This item is not returnable.
View return policy
Description
Kallikrein 5 Polyclonal specifically detects Kallikrein 5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| Kallikrein 5 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.21, EC 3.4.21.4, kallikrein 5, Kallikrein-like protein 2, kallikrein-related peptidase 5, KLK5, KLKL2, KLK-L2, KLK-L2EC 3.4.21.-, SCTE, SCTEkallikrein-5, Stratum corneum tryptic enzyme | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 25818 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q9Y337 | |
| KLK5 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 5 Ile67-Ser293 Accession # Q9Y337 | |
| 25 μg | |
| Primary | |
| Detects human Kallikrein 5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human (rh) KLK8 and rhKLK13 is observed and less than 1% cross-reactivity with rhKLK1, -3, -4, -7, -9, -10, -11, and -12 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction